Product Details

SNP ID
rs728212
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:158291600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATTTTTGGAATTTTTTTTCTTG[A/T]GCTTTGTTATCTTCCACCTGTGGTC
Phenotype
MIM: 188360 MIM: 188340
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CD1B PubMed Links

Gene Details

Gene
CD1B
Gene Name
CD1b molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001764.2 Intron NP_001755.1
XM_011510119.2 Intron XP_011508421.1
XM_017002784.1 Intron XP_016858273.1
XM_017002785.1 Intron XP_016858274.1
XM_017002786.1 Intron XP_016858275.1
Gene
CD1C
Gene Name
CD1c molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001765.2 Intron NP_001756.2
XM_005245579.4 Intron XP_005245636.1

View Full Product Details