Product Details

SNP ID
rs1043768
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:153974556 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCTCATTAATGATCTGAAAGAA[A/G]AAGATGGGGAAAAGGGGATTCCACC
Phenotype
MIM: 607138 MIM: 604671 MIM: 602672 MIM: 604740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREB3L4 PubMed Links

Gene Details

Gene
CREB3L4
Gene Name
cAMP responsive element binding protein 3 like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001255978.1 1287 Intron NP_001242907.1
NM_001255979.1 1287 Intron NP_001242908.1
NM_001255980.1 1287 Intron NP_001242909.1
NM_001255981.1 1287 Intron NP_001242910.1
NM_130898.3 1287 Intron NP_570968.1
XM_006711172.2 1287 Intron XP_006711235.1
XM_017000372.1 1287 Intron XP_016855861.1
Gene
JTB
Gene Name
jumping translocation breakpoint
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006694.3 1287 UTR 3 NP_006685.1
Gene
RAB13
Gene Name
RAB13, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
SLC39A1
Gene Name
solute carrier family 39 member 1
There are no transcripts associated with this gene.

View Full Product Details