Product Details

SNP ID
rs1511050
Assay Type
Validated
NCBI dbSNP Submissions
36
Location
Chr.1:214613318 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCAGCTAACTATGCCACCTCCTACC[C/T]TTTTCTGGCCTTCTTCGAGTTCTTC
Phenotype
MIM: 600236
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPF PubMed Links

Gene Details

Gene
CENPF
Gene Name
centromere protein F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016343.3 Intron NP_057427.3
XM_011509082.2 Intron XP_011507384.1
XM_017000086.1 Intron XP_016855575.1

View Full Product Details