Product Details

SNP ID
rs3843707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:12902931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGACTGAGGCGGACTTGAGACAC[A/G]TAGCCTGGGAGCCCGTGGCCCCGAA
Phenotype
MIM: 613346
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHISA9 PubMed Links

Gene Details

Gene
SHISA9
Gene Name
shisa family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145204.2 Intron NP_001138676.2
NM_001145205.1 Intron NP_001138677.2
XM_005255539.3 Intron XP_005255596.2
XM_011522642.2 Intron XP_011520944.1

View Full Product Details