Product Details

SNP ID
rs151181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28479196 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATACTTTTCGTATATGTAAGAAT[C/T]GCCTGGGGATCTAGGTACAAATGCC
Phenotype
MIM: 607042
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLN3 PubMed Links

Gene Details

Gene
CLN3
Gene Name
CLN3, battenin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000086.2 Intron NP_000077.1
NM_001042432.1 Intron NP_001035897.1
NM_001286104.1 Intron NP_001273033.1
NM_001286105.1 Intron NP_001273034.1
NM_001286109.1 Intron NP_001273038.1
NM_001286110.1 Intron NP_001273039.1

View Full Product Details