Product Details

SNP ID
hCV7489395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153691525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCACCTGTGACCAGCTTGCTGAC[C/T]GCCGGTCCCCTGTCATCGAGTTCTG
Phenotype
MIM: 300398 MIM: 300680 MIM: 300036
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCAP31 PubMed Links

Gene Details

Gene
BCAP31
Gene Name
B-cell receptor-associated protein 31
There are no transcripts associated with this gene.

Gene
PNCK
Gene Name
pregnancy up-regulated nonubiquitous CaM kinase
There are no transcripts associated with this gene.

Gene
SLC6A8
Gene Name
solute carrier family 6 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142805.1 443 Intron NP_001136277.1
NM_001142806.1 443 Missense Mutation CGC,TGC R91C NP_001136278.1
NM_005629.3 443 Intron NP_005620.1

View Full Product Details