Product Details

SNP ID
rs12877
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:154401914 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCAGCCAGTGTTTTTCCTGACCTCT[C/G]TTCTTTGGAGAGGAGGATGGAAGGG
Phenotype
MIM: 300081 MIM: 312173 MIM: 300394
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNASE1L1 PubMed Links

Gene Details

Gene
DNASE1L1
Gene Name
deoxyribonuclease I-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009932.2 2333 UTR 3 NP_001009932.1
NM_001009933.2 2333 UTR 3 NP_001009933.1
NM_001009934.2 2333 UTR 3 NP_001009934.1
NM_001303620.1 2333 UTR 3 NP_001290549.1
NM_006730.3 2333 UTR 3 NP_006721.1
XM_005277829.4 2333 UTR 3 XP_005277886.1
XM_011531122.2 2333 UTR 3 XP_011529424.1
XM_017029332.1 2333 UTR 3 XP_016884821.1
Gene
LOC107987332
Gene Name
uncharacterized LOC107987332
There are no transcripts associated with this gene.

Gene
RPL10
Gene Name
ribosomal protein L10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256577.2 2333 UTR 3 NP_001243506.2
NM_001256580.2 2333 UTR 3 NP_001243509.2
NM_001303624.1 2333 UTR 3 NP_001290553.1
NM_001303625.1 2333 UTR 3 NP_001290554.1
NM_001303626.1 2333 UTR 3 NP_001290555.1
NM_006013.4 2333 UTR 3 NP_006004.3
Gene
SNORA70
Gene Name
small nucleolar RNA, H/ACA box 70
There are no transcripts associated with this gene.

Gene
TAZ
Gene Name
tafazzin
There are no transcripts associated with this gene.

View Full Product Details