Product Details

SNP ID
rs1001046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17515548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCACAGGTGCTGGGCCTGGGCAA[A/G]CCTGGCCCCCACTCCCTGTAGTGGG
Phenotype
MIM: 609967 MIM: 604951 MIM: 600691
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM129C PubMed Links

Gene Details

Gene
FAM129C
Gene Name
family with sequence similarity 129 member C
There are no transcripts associated with this gene.

Gene
PGLS
Gene Name
6-phosphogluconolactonase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012088.2 Intron NP_036220.1
XM_011527897.2 Intron XP_011526199.1
Gene
SLC27A1
Gene Name
solute carrier family 27 member 1
There are no transcripts associated with this gene.

View Full Product Details