Product Details

SNP ID
rs1064796
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.19:15913852 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
TCGGCAGGATGCGGAAGTGCAGCAG[C/G]GTGAGCGCCAGGACCACCTTCATCT
Phenotype
MIM: 611517
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYP4F11 PubMed Links

Gene Details

Gene
CYP4F11
Gene Name
cytochrome P450 family 4 subfamily F member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128932.1 1913 Silent Mutation ACC,ACG T485T NP_001122404.1
NM_021187.3 1913 Silent Mutation ACC,ACG T485T NP_067010.3

View Full Product Details