Product Details

SNP ID
rs888668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18429806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACGGTGGGCTTAGGGGGCTCCCG[C/G]CTCCAGCCCCTCCCCCAGTTTAGGA
Phenotype
MIM: 611670 MIM: 607391
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ISYNA1 PubMed Links

Gene Details

Gene
ISYNA1
Gene Name
inositol-3-phosphate synthase 1
There are no transcripts associated with this gene.

Gene
SSBP4
Gene Name
single stranded DNA binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009998.3 Intron NP_001009998.1
NM_032627.4 Intron NP_116016.1
XM_005259790.3 Intron XP_005259847.1
XM_006722665.3 Intron XP_006722728.1
XM_006722666.2 Intron XP_006722729.1
XM_006722668.2 Intron XP_006722731.1
XM_017026431.1 Intron XP_016881920.1
XM_017026432.1 Intron XP_016881921.1
XM_017026433.1 Intron XP_016881922.1
XM_017026434.1 Intron XP_016881923.1
XM_017026435.1 Intron XP_016881924.1
XM_017026436.1 Intron XP_016881925.1
XM_017026437.1 Intron XP_016881926.1

View Full Product Details