Product Details

SNP ID
rs985315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:21251246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCTTTCTTTTCACTCCAACA[A/T]TGTCACTCATCTATTTATGTTGTTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GREB1L PubMed Links

Gene Details

Gene
GREB1L
Gene Name
growth regulation by estrogen in breast cancer 1 like
There are no transcripts associated with this gene.

View Full Product Details