Product Details

SNP ID
rs838890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:124782955 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAACAACCCTCAACGATTGCAGGT[A/G]GCTGAGATGTGGGGAGCTGCCGCTC
Phenotype
MIM: 601040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCARB1 PubMed Links

Gene Details

Gene
SCARB1
Gene Name
scavenger receptor class B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082959.1 Intron NP_001076428.1
NM_005505.4 Intron NP_005496.4

View Full Product Details