Product Details

SNP ID
rs1770987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104088984 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAGTAATACAAGTTACAAAATC[A/G]TAGTTAGGTGTGAAAGCCGGTGTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASPG PubMed Links

Gene Details

Gene
ASPG
Gene Name
asparaginase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080464.2 Intron NP_001073933.2
XM_005267590.4 Intron XP_005267647.1
XM_017021265.1 Intron XP_016876754.1
XM_017021266.1 Intron XP_016876755.1
XM_017021267.1 Intron XP_016876756.1
XM_017021268.1 Intron XP_016876757.1
XM_017021269.1 Intron XP_016876758.1
XM_017021270.1 Intron XP_016876759.1
XM_017021271.1 Intron XP_016876760.1
XM_017021272.1 Intron XP_016876761.1
XM_017021273.1 Intron XP_016876762.1
XM_017021274.1 Intron XP_016876763.1
XM_017021275.1 Intron XP_016876764.1
XM_017021276.1 Intron XP_016876765.1
Gene
LOC105370691
Gene Name
uncharacterized LOC105370691
There are no transcripts associated with this gene.

View Full Product Details