Product Details
- SNP ID
-
rs7534
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:95464530 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATAAATATAGTTTAAAAAAATGTTG[C/T]TTTGGGTAGAAGTCTTGATTCATTG
- Phenotype
-
MIM: 614589
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TTC37
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201040714] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TTC37
- Gene Name
- tetratricopeptide repeat domain 37
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014639.3 |
5069 |
UTR 3 |
|
|
NP_055454.1 |
View Full Product Details