Product Details

SNP ID
rs3915490
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62213393 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCTGTGTTGTAAAAGTGGTATCA[C/G]AAACAGTCCAAATGATCAGGTCTTC
Phenotype
MIM: 604398
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
SCGB2A1 PubMed Links

Gene Details

Gene
SCGB2A1
Gene Name
secretoglobin family 2A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002407.2 Intron NP_002398.1

View Full Product Details