Product Details

SNP ID
rs3913486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:36153016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGACTTTGTGATTGTCCGCAGGC[C/T]TAAGCTAAATCGAGAGAACTTTCCA
Phenotype
MIM: 601436
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMBRD2 PubMed Links

Gene Details

Gene
LMBRD2
Gene Name
LMBR1 domain containing 2
There are no transcripts associated with this gene.

Gene
MIR580
Gene Name
microRNA 580
There are no transcripts associated with this gene.

Gene
SKP2
Gene Name
S-phase kinase-associated protein 2, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243120.1 474 UTR 5 NP_001230049.1
NM_005983.3 474 Missense Mutation CCT,CTT P85L NP_005974.2
NM_032637.3 474 Missense Mutation CCT,CTT P85L NP_116026.1

View Full Product Details