Product Details

SNP ID
rs805295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31707520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAGAGCTGCCATGTCCCTCACCA[T/C]CTACTCTACATGGGGACGAACACCT
Phenotype
MIM: 142620 MIM: 606038 MIM: 610437 MIM: 611404
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ABHD16A PubMed Links

Gene Details

Gene
ABHD16A
Gene Name
abhydrolase domain containing 16A
There are no transcripts associated with this gene.

Gene
LY6G6D
Gene Name
lymphocyte antigen 6 complex, locus G6D
There are no transcripts associated with this gene.

Gene
LY6G6E
Gene Name
lymphocyte antigen 6 complex, locus G6E (pseudogene)
There are no transcripts associated with this gene.

Gene
LY6G6F
Gene Name
lymphocyte antigen 6 complex, locus G6F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003693.1 115 Missense Mutation CCT,TCT P39S NP_001003693.1
Gene
MIR4646
Gene Name
microRNA 4646
There are no transcripts associated with this gene.

View Full Product Details