Product Details

SNP ID
rs1064202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47352883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTATCATCTGTGGTGGTCCTGT[C/G]CCGTGACCAGGAGGAAAAATTAGCT
Phenotype
MIM: 615475
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DHX34 PubMed Links

Gene Details

Gene
DHX34
Gene Name
DEAH-box helicase 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014681.5 151 UTR 5 NP_055496.2
XM_005259443.3 151 UTR 5 XP_005259500.1
XM_011527550.2 151 UTR 5 XP_011525852.1
XM_011527551.2 151 UTR 5 XP_011525853.1
XM_017027524.1 151 UTR 5 XP_016883013.1

View Full Product Details