Product Details

SNP ID
rs1363730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:163504008 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAGCAGCCAAGTTAAATTCTTT[C/T]TGTTCCTCAAGACTAGGCTGCTCTG
Phenotype
MIM: 605527
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMMR-AS1 PubMed Links

Gene Details

Gene
HMMR-AS1
Gene Name
HMMR antisense RNA 1
There are no transcripts associated with this gene.

Gene
MAT2B
Gene Name
methionine adenosyltransferase 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013283.4 Intron NP_037415.1
NM_182796.2 Intron NP_877725.1

View Full Product Details