Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001171506.2 | Intron | NP_001164977.1 | |||
NM_001171507.2 | Intron | NP_001164978.1 | |||
NM_001171508.2 | Intron | NP_001164979.1 | |||
NM_001171509.2 | Intron | NP_001164980.1 | |||
NM_001171510.2 | Intron | NP_001164981.1 | |||
NM_001171511.2 | Intron | NP_001164982.1 | |||
NM_139279.5 | Intron | NP_644808.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001288951.1 | Intron | NP_001275880.1 | |||
NM_001288953.1 | Intron | NP_001275882.1 | |||
NM_001288955.1 | Intron | NP_001275884.1 | |||
NM_020458.3 | Intron | NP_065191.2 | |||
XM_005264439.3 | Intron | XP_005264496.1 | |||
XM_011532998.2 | Intron | XP_011531300.1 | |||
XM_011532999.2 | Intron | XP_011531301.1 | |||
XM_011533000.2 | Intron | XP_011531302.1 | |||
XM_011533001.2 | Intron | XP_011531303.1 | |||
XM_017004524.1 | Intron | XP_016860013.1 | |||
XM_017004525.1 | Intron | XP_016860014.1 | |||
XM_017004526.1 | Intron | XP_016860015.1 | |||
XM_017004527.1 | Intron | XP_016860016.1 | |||
XM_017004528.1 | Intron | XP_016860017.1 | |||
XM_017004529.1 | Intron | XP_016860018.1 | |||
XM_017004530.1 | Intron | XP_016860019.1 |