Product Details

SNP ID
rs13178
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:36796985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGTCCTGCCTCAGACAGGCTC[A/G]GATGGAGATGAGGTCCGTTGGCCAT
Phenotype
MIM: 138243
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIK3 PubMed Links

Gene Details

Gene
GRIK3
Gene Name
glutamate ionotropic receptor kainate type subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000831.3 7761 UTR 3 NP_000822.2

View Full Product Details