Product Details

SNP ID
rs1792765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:105096065 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGCTATTCAACGTTTTAAAGATT[C/T]CATTGAGTTGTAATACATCTGGGAC
Phenotype
MIM: 609490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD17 PubMed Links

Gene Details

Gene
CARD17
Gene Name
caspase recruitment domain family member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007232.1 Intron NP_001007233.1
XM_011542838.2 Intron XP_011541140.1

View Full Product Details