Product Details

SNP ID
rs1065727
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:225920091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTCCTCACCACTTTGCCTTGG[C/G]ATCACACCAACCCTGCCTCGGGCTT
Phenotype
MIM: 616406
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6741 PubMed Links

Gene Details

Gene
MIR6741
Gene Name
microRNA 6741
There are no transcripts associated with this gene.

Gene
PYCR2
Gene Name
pyrroline-5-carboxylate reductase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271681.1 1335 UTR 3 NP_001258610.1
NM_013328.3 1335 UTR 3 NP_037460.2

View Full Product Details