Product Details

SNP ID
rs1281022
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:10946567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAAATCCCACTTCCAGTGATGG[A/G]TCTGCCTGGAGGTGGGCGTCTGGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf127 PubMed Links

Gene Details

Gene
C1orf127
Gene Name
chromosome 1 open reading frame 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170754.1 2585 UTR 3 NP_001164225.1
XM_017000373.1 2585 UTR 3 XP_016855862.1

View Full Product Details