Product Details

SNP ID
rs1347400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:191334194 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCACAATAAAAAAAAATTTTCC[A/G]TGGTACATGAACATGATCTTTGCTG
Phenotype
MIM: 611051
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC50 PubMed Links

Gene Details

Gene
CCDC50
Gene Name
coiled-coil domain containing 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174908.3 Intron NP_777568.1
NM_178335.2 Intron NP_848018.1
XM_011512460.1 Intron XP_011510762.1
Gene
UTS2B
Gene Name
urotensin 2B
There are no transcripts associated with this gene.

View Full Product Details