Product Details

SNP ID
rs1248860
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:84966628 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTATGAAACTGAGTGATAAGAACTT[A/G]TTACTACTTGAATAGATACAGAGCT
Phenotype
MIM: 609938
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CADM2 PubMed Links

Gene Details

Gene
CADM2
Gene Name
cell adhesion molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167674.1 Intron NP_001161146.1
NM_001167675.1 Intron NP_001161147.1
NM_001256502.1 Intron NP_001243431.1
NM_001256503.1 Intron NP_001243432.1
NM_001256504.1 Intron NP_001243433.1
NM_001256505.1 Intron NP_001243434.1
NM_153184.3 Intron NP_694854.2
XM_006713081.3 Intron XP_006713144.1
XM_006713082.3 Intron XP_006713145.1
XM_017006062.1 Intron XP_016861551.1
XM_017006063.1 Intron XP_016861552.1

View Full Product Details