Product Details

SNP ID
rs1063157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:146071339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCAATGAACTCCCGGATAAAAT[C/G]AAGCCATACATTCTCCAGATCAACT
Phenotype
MIM: 601865
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PLOD2 PubMed Links

Gene Details

Gene
PLOD2
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000935.2 1996 Missense Mutation CAT,GAT H624D NP_000926.2
NM_182943.2 1996 Missense Mutation CAT,GAT H645D NP_891988.1
XM_005247535.3 1996 Missense Mutation CAT,GAT H553D XP_005247592.1
XM_017006625.1 1996 Missense Mutation CAT,GAT H553D XP_016862114.1

View Full Product Details