Product Details

SNP ID
rs1359109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:48657977 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTCTTTTAAGTAAGCTGGTTTCT[C/T]TTTATTTTGAGTCTTACCTTAAAAA
Phenotype
MIM: 605666
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYSLTR2 PubMed Links

Gene Details

Gene
CYSLTR2
Gene Name
cysteinyl leukotriene receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308465.1 Intron NP_001295394.1
NM_001308467.1 Intron NP_001295396.1
NM_001308468.1 Intron NP_001295397.1
NM_001308469.1 Intron NP_001295398.1
NM_001308470.1 Intron NP_001295399.1
NM_001308471.1 Intron NP_001295400.1
NM_001308476.1 Intron NP_001295405.1
NM_020377.3 Intron NP_065110.1

View Full Product Details