Product Details
- SNP ID
-
rs1514403
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70239296 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCACCAAGCCAAGAGCTGCTACAA[C/T]ATTGAGTGGGAAGAATATCTAACAG
- Phenotype
-
MIM: 601695
MIM: 607241
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CSN3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75721346] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CSN3
- Gene Name
- casein kappa
- Gene
- FDCSP
- Gene Name
- follicular dendritic cell secreted protein
There are no transcripts associated with this gene.
View Full Product Details