Product Details

SNP ID
rs1230399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43768035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACAAAAGTCCCTGATATTTCTTT[C/T]GTTTTAGGTAAGACTCCTTCACAAG
Phenotype
MIM: 600183 MIM: 605740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP3 PubMed Links

Gene Details

Gene
DUSP3
Gene Name
dual specificity phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004090.3 2208 UTR 3 NP_004081.1
Gene
SOST
Gene Name
sclerostin
There are no transcripts associated with this gene.

View Full Product Details