Product Details

SNP ID
rs11713
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43768456 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAAAGGATCCATTTGGACACTCC[A/G]CCATGTCTCATGTCTGCCTGGCTGA
Phenotype
MIM: 600183 MIM: 605740
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C17orf105 PubMed Links

Gene Details

Gene
C17orf105
Gene Name
chromosome 17 open reading frame 105
There are no transcripts associated with this gene.

Gene
DUSP3
Gene Name
dual specificity phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004090.3 1787 UTR 3 NP_004081.1
Gene
SOST
Gene Name
sclerostin
There are no transcripts associated with this gene.

View Full Product Details