Product Details

SNP ID
rs1800644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:90060825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCATACTTGGCAGATAATCTGT[A/T]ATGTCAGGCCACCCAAGTTTTTAGC
Phenotype
MIM: 114050 MIM: 222745
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CALB1 PubMed Links

Gene Details

Gene
CALB1
Gene Name
calbindin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004929.3 Intron NP_004920.1
Gene
DECR1
Gene Name
2,4-dienoyl-CoA reductase 1, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details