Product Details
- SNP ID
-
rs1033740
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:36475809 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAATAGAGACCTAAAAGAATATAT[A/G]GGGCTTTTAGTCTCACTTTAAAGTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SFTA3
PubMed Links
Gene Details
- Gene
- SFTA3
- Gene Name
- surfactant associated 3
View Full Product Details