Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001098518.1 | Intron | NP_001091988.1 | |||
NM_015234.4 | Intron | NP_056049.4 | |||
XM_005248892.2 | Intron | XP_005248949.1 | |||
XM_005248893.3 | Intron | XP_005248950.1 | |||
XM_005248894.3 | Intron | XP_005248951.1 | |||
XM_005248895.3 | Intron | XP_005248952.1 | |||
XM_011514353.2 | Intron | XP_011512655.1 | |||
XM_011514354.2 | Intron | XP_011512656.1 | |||
XM_017010418.1 | Intron | XP_016865907.1 | |||
XM_017010419.1 | Intron | XP_016865908.1 | |||
XM_017010420.1 | Intron | XP_016865909.1 | |||
XM_017010421.1 | Intron | XP_016865910.1 | |||
XM_017010422.1 | Intron | XP_016865911.1 | |||
XM_017010423.1 | Intron | XP_016865912.1 | |||
XM_017010424.1 | Intron | XP_016865913.1 |