Product Details
- SNP ID
-
rs1475696
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:97929115 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCGTTAATTAATTGACCATATCGA[C/T]CTTGCTGGGTTTAGAATGAAGCAGT
- Phenotype
-
MIM: 610715
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HEMGN
PubMed Links
Gene Details
- Gene
- HEMGN
- Gene Name
- hemogen
- Gene
- TRMO
- Gene Name
- tRNA methyltransferase O
There are no transcripts associated with this gene.
View Full Product Details