Product Details

SNP ID
rs1024516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:86651882 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCACTACCTTCCTTGGTTCATA[A/C]AATTTTCCAAATTTTAAACTGAAGT
Phenotype
MIM: 601115
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GRM3 PubMed Links

Gene Details

Gene
GRM3
Gene Name
glutamate metabotropic receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000840.2 Intron NP_000831.2
XM_011516088.1 Intron XP_011514390.1
XM_017012073.1 Intron XP_016867562.1

View Full Product Details