Product Details

SNP ID
rs1451525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:137387625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGCATGAACGGATGGGGTATTT[C/T]TGACATCACATACAAGCAACAGTGT
Phenotype
MIM: 604072
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGKI PubMed Links

Gene Details

Gene
DGKI
Gene Name
diacylglycerol kinase iota
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321708.1 6519 Intron NP_001308637.1
NM_001321709.1 6519 Intron NP_001308638.1
NM_001321710.1 6519 Intron NP_001308639.1
NM_004717.3 6519 Intron NP_004708.1
XM_011516682.2 6519 Intron XP_011514984.2
XM_011516684.2 6519 UTR 3 XP_011514986.2
XM_017012780.1 6519 UTR 3 XP_016868269.1
XM_017012781.1 6519 UTR 3 XP_016868270.1
XM_017012782.1 6519 UTR 3 XP_016868271.1
XM_017012783.1 6519 UTR 3 XP_016868272.1
XM_017012784.1 6519 UTR 3 XP_016868273.1
XM_017012785.1 6519 UTR 3 XP_016868274.1
XM_017012786.1 6519 UTR 3 XP_016868275.1
XM_017012787.1 6519 UTR 3 XP_016868276.1
XM_017012788.1 6519 UTR 3 XP_016868277.1
XM_017012789.1 6519 UTR 3 XP_016868278.1

View Full Product Details