Product Details

SNP ID
rs2762
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:96937594 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTTGTAAGTGACTATTCCAATGTT[C/T]ATTCTTTGTTTTTGGAAGAATGCTT
Phenotype
MIM: 606832 MIM: 151300
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERAP1 PubMed Links

Gene Details

Gene
ERAP1
Gene Name
endoplasmic reticulum aminopeptidase 1
There are no transcripts associated with this gene.

Gene
LNPEP
Gene Name
leucyl and cystinyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005575.2 Intron NP_005566.2
NM_175920.3 Intron NP_787116.2
Gene
LOC101929747
Gene Name
uncharacterized LOC101929747
There are no transcripts associated with this gene.

View Full Product Details