Product Details

SNP ID
rs1800545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:111077780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTAGGAGCTCGGAGCAAGAAGGC[A/G]CCCACCGAGAGCGTCTGAAGCGCGA
Phenotype
MIM: 104210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADRA2A PubMed Links

Gene Details

Gene
ADRA2A
Gene Name
adrenoceptor alpha 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000681.3 749 UTR 5 NP_000672.3

View Full Product Details