Product Details

SNP ID
rs1494318
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:110877821 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGCTTTGGCTTCGTTTTATTTTAC[C/T]CCTACTTCTTCCTCTACAACTGCTT
Phenotype
MIM: 151525 MIM: 615354
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD53 PubMed Links

Gene Details

Gene
CD53
Gene Name
CD53 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000560.3 Intron NP_000551.1
NM_001040033.1 Intron NP_001035122.1
NM_001320638.1 Intron NP_001307567.1
Gene
LRIF1
Gene Name
ligand dependent nuclear receptor interacting factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006945.1 Intron NP_001006946.1
NM_018372.3 Intron NP_060842.3
XM_005271029.3 Intron XP_005271086.1
XM_011541768.1 Intron XP_011540070.1
XM_017001768.1 Intron XP_016857257.1
XM_017001769.1 Intron XP_016857258.1

View Full Product Details