Product Details

SNP ID
rs822436
Assay Type
Validated
NCBI dbSNP Submissions
52
Location
Chr.1:156922602 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCTGTGCTGTGTGACAGGTAACTA[C/T]CTGCTAAAGTGCAGAGAGCCCATTT
Phenotype
MIM: 610278
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC71 PubMed Links

Gene Details

Gene
LRRC71
Gene Name
leucine rich repeat containing 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144702.2 Intron NP_653303.2
XM_005244926.3 Intron XP_005244983.1
XM_005244927.3 Intron XP_005244984.1
XM_005244928.2 Intron XP_005244985.1
XM_006711185.3 Intron XP_006711248.1
XM_006711186.3 Intron XP_006711249.1
XM_006711187.3 Intron XP_006711250.1
XM_011509239.2 Intron XP_011507541.1
XM_011509240.2 Intron XP_011507542.1
XM_011509241.2 Intron XP_011507543.1
XM_017000459.1 Intron XP_016855948.1
XM_017000460.1 Intron XP_016855949.1
XM_017000461.1 Intron XP_016855950.1
XM_017000462.1 Intron XP_016855951.1
Gene
PEAR1
Gene Name
platelet endothelial aggregation receptor 1
There are no transcripts associated with this gene.

View Full Product Details