Product Details

SNP ID
rs1058663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101626485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTTAATGCTGGAAAGAAAAAAA[A/G]AAGAAAGAAAGAAACCATCCCTGTC
Phenotype
MIM: 300364
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMCX3 PubMed Links

Gene Details

Gene
ARMCX3
Gene Name
armadillo repeat containing, X-linked 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016607.3 2168 UTR 3 NP_057691.1
NM_177947.2 2168 UTR 3 NP_808816.1
NM_177948.2 2168 UTR 3 NP_808817.1
XM_005262141.2 2168 UTR 3 XP_005262198.1
Gene
ARMCX6
Gene Name
armadillo repeat containing, X-linked 6
There are no transcripts associated with this gene.

View Full Product Details