Product Details
- SNP ID
-
hCV7622182
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:4189382 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTTGTGTTGTTCACGGAACCTAC[A/G]CACTGTTCCCTGCTTCTCAGACTCT
- Phenotype
-
MIM: 607806
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OTOP1
PubMed Links
Gene Details
- Gene
- OTOP1
- Gene Name
- otopetrin 1
There are no transcripts associated with this gene.
View Full Product Details