Product Details
- SNP ID
-
rs2043508
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:11369897 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAATAGACACCTGTACTATTAGTT[A/G]CAGGTGTTTCAGATTTGTTAATCTA
- Phenotype
-
MIM: 615174
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM167A-AS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs374633085] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM167A-AS1
- Gene Name
- FAM167A antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- TDH
- Gene Name
- L-threonine dehydrogenase (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details