Product Details

SNP ID
rs2736287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11370497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGTATATGTGATACCTTGTCT[A/G]TTGATCCATTTGTCTGAAGTCCTTA
Phenotype
MIM: 615174
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM167A-AS1 PubMed Links

Gene Details

Gene
FAM167A-AS1
Gene Name
FAM167A antisense RNA 1
There are no transcripts associated with this gene.

Gene
TDH
Gene Name
L-threonine dehydrogenase (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details