Product Details
- SNP ID
-
rs1800883
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:155070881 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACACCCCTTCTGCAAGTACCCCAGG[G/C]CGGTCTCCTGACCCAGAGATGGATT
- Phenotype
-
MIM: 601305
- Polymorphism
- G/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HTR5A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs55967309] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HTR5A
- Gene Name
- 5-hydroxytryptamine receptor 5A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024012.3 |
558 |
UTR 5 |
|
|
NP_076917.1 |
- Gene
- HTR5A-AS1
- Gene Name
- HTR5A antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details