Product Details

SNP ID
rs7096899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:50245117 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAGCAAGGTCAATTAAAAAGCTA[A/G]AAGAAGATAATGATGATGTTGTAAT
Phenotype
MIM: 611202
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASAH2 PubMed Links

Gene Details

Gene
ASAH2
Gene Name
N-acylsphingosine amidohydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143974.1 Intron NP_001137446.1
NM_019893.2 Intron NP_063946.2
XM_011539970.2 Intron XP_011538272.1
XM_011539971.2 Intron XP_011538273.1
XM_011539972.2 Intron XP_011538274.1
XM_011539973.2 Intron XP_011538275.1
XM_017016435.1 Intron XP_016871924.1
XM_017016436.1 Intron XP_016871925.1

View Full Product Details