Product Details

SNP ID
rs4380146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46603377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTAGAGGTGGGACAAGTCACTG[G/T]TAACTGGCCTTGGACTTTGGCCTGA
Phenotype
MIM: 114183 MIM: 600022
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CALM3 PubMed Links

Gene Details

Gene
CALM3
Gene Name
calmodulin 3
There are no transcripts associated with this gene.

Gene
PPP5D1
Gene Name
PPP5 tetratricopeptide repeat domain containing 1
There are no transcripts associated with this gene.

Gene
PTGIR
Gene Name
prostaglandin I2 (prostacyclin) receptor (IP)
There are no transcripts associated with this gene.

View Full Product Details