Product Details

SNP ID
rs2919847
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:44701052 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGTGCAGTCACCTTTTACTGATTTT[A/C]TTCCTTGGTGACTCCCAGGCTCTCT
Phenotype
MIM: 614591
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM16 PubMed Links

Gene Details

Gene
CEACAM16
Gene Name
carcinoembryonic antigen related cell adhesion molecule 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039213.3 497 Intron NP_001034302.2
XM_017026795.1 497 UTR 5 XP_016882284.1

View Full Product Details