Product Details
- SNP ID
-
rs1555897
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:807734 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGCTCCGAGACATCACACACTTGG[A/C]TATGGCCTGGGCTGACATAAGTTCC
- Phenotype
-
MIM: 616513
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LARP4B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs146171221] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LARP4B
- Gene Name
- La ribonucleoprotein domain family member 4B
View Full Product Details